1,3-Cyclohexadiene, 96%, stabilized
1,3-Cyclohexadiene, 96%, stabilized
1,3-Cyclohexadiene, 96%, stabilized
Acros Organics

1,3-Cyclohexadiene, 96%, stabilized

CAS: 592-57-4 | C6H8 | 80.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number 111100250
also known as 11110
HSN 29021900
Price (INR)
28,406.00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
28,406.00
Each
Chemical Identifiers
CAS592-57-4
IUPAC Namecyclohexa-1,3-diene
Molecular FormulaC6H8
InChI KeyMGNZXYYWBUKAII-UHFFFAOYSA-N
SMILESC1CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer0.1 % BHT
Appearance (Form)Liquid
Refractive index1.4715 to 1.4765 (20°C, nm)
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only