1,2-Cyclohexanedione, 98%
1,2-Cyclohexanedione, 98%
1,2-Cyclohexanedione, 98%
1,2-Cyclohexanedione, 98%
Acros Organics

1,2-Cyclohexanedione, 98%

CAS: 765-87-7 | C6H8O2 | 112.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
111160100
also known as 11116
10 gGlass bottle
Catalog number 111160100
also known as 11116
HSN 29142200
Price (INR)
14,061.00
Each
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,061.00
Each
Add to cart
Chemical Identifiers
CAS765-87-7
IUPAC Namecyclohexane-1,2-dione
Molecular FormulaC6H8O2
InChI KeyOILAIQUEIWYQPH-UHFFFAOYSA-N
SMILESO=C1CCCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5180 to 1.5200 (20°C, 589 nm)
Appearance (Form)Liquid after melting
GC>=97.5 %
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only