Cyclohexyl chloride, 98%
Cyclohexyl chloride, 98%
Cyclohexyl chloride, 98%
Acros Organics

Cyclohexyl chloride, 98%

CAS: 542-18-7 | C6H11Cl | 118.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 111301000
also known as 11130
HSN 29035900
Price (INR)
3,503.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,503.00
Each
Chemical Identifiers
CAS542-18-7
IUPAC Namechlorocyclohexane
Molecular FormulaC6H11Cl
InChI KeyUNFUYWDGSFDHCW-UHFFFAOYSA-N
SMILESClC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4595 to 1.4645 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only