Product Image
Thermo Scientific™
Desolvating Systems
Improve sensitivity and signal stability by adding one of these Thermo Scientific™ ESI™ or CETAC™ Desolvating Systems to the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1114021Aridus II Desolvating System
1299560Apex Q Desolvating System
Catalog number 1114021
Price (INR)
-
Type:
Aridus II Desolvating System
Improve sensitivity and signal stability, by adding a specialized Thermo Scientific™ Desolvating System accessory such as these options from Thermo Scientific™ ESI™ and Thermo Scientific™ CETAC™, compatible with the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
Gain Performance Improvements with Advanced Accessories
CETAC Aridus II™ Desolvating Nebulizer System (with membrane desolvation)
The Aridus II Desolvating System offers sensitivity enhancement of up to 10x for ICP-MS and improved signal stability, even for organic solvents. A unique PTFE membrane desolvator yields low oxide and hydride levels, while the modular design ensures easy maintenance.
ESI APEX Q Desolvating Nebulizer System (with membrane desolvation upgrade option)
The Apex Q Desolvating System has an O-ring-free quartz flow path for high sensitivity and low background for samples that do not contain hydrofluoric acid. The Apex Q system provides an ultra-fast rinse-out for high sensitivity ICP introduction systems.
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
TypeAridus II Desolvating System
Unit SizeEach