RP4 Windshielded Swinging-Bucket Rotor
RP4 Windshielded Swinging-Bucket Rotor
Actual product may vary
Thermo Scientific™
RP4 Windshielded Swinging-Bucket Rotor
Choose from unsealed rectangular or round buckets for capacity flexibility with the Thermo Scientific™ RP4 Windshielded Swinging-Bucket Rotor, a large volume rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
11178143Unsealed Round Bucket
11178073Rotor Lid
11178088Unsealed Rectangular Bucket
11178250RP4 Windshielded Swinging Bucket Rotor
Catalog number 11178143
Price (INR)
-
Request A Quote
Description:
Unsealed Round Bucket
Choose from unsealed rectangular or round buckets for capacity flexibility with the Thermo Scientific™ RP4 Windshielded Swinging-Bucket Rotor, a large volume rotor.
Ordering Information:
Order rotor body (11178250) and buckets and inserts to complete assembly. Order 11178088 for unsealed rectangular bucket (set of 4) and 11178143 for unsealed round bucket (set of 4). Contact your Sales Representative for information.
Compatible with:
Thermo Scientific KR4i and Sorvall HT6 Centrifuges
Recommended for:
Pharmaceutical, clinical, genomics, proteomics, biochemistry, pathology research, RIA and large-volume processing
Specifications
DescriptionUnsealed Round Bucket
For Use With (Equipment)Thermo Scientific KR4i Large Capacity Centrifuges
K Factor12,994/14,669 (w/o lid)
Min. RCF2,180/1,931 x g (w/o lid)
Radius (Metric)273 mm
Radius (Metric) Min.108 mm
Capacity4 x 1600 mL
MaterialAluminum
Max. RCF5,372 x g
Max. Speed4,000 rpm
TypeBucket Rotor
Unit SizeEach