F-G3 Fixed-Angle Rotor
Thermo Scientific™
F-G3 Fixed-Angle Rotor
Integrate the Thermo Scientific™ F-G3 Fixed-Angle Rotor with the Thermo Scientific™ CL10 centrifuge.
Have Questions?
Catalog NumberCapacity
1121001924 x 15 mL
Catalog number 11210019
Price (INR)
-
Capacity:
24 x 15 mL
Integrate the Thermo Scientific™ F-G3 Fixed-Angle Rotor with the Thermo Scientific™ CL10 centrifuge.
Specifications
AngleFixed
DescriptionF-G3 Fixed-Angle Rotor
For Use With (Equipment)Thermo Scientific CL 10 Centrifuges
Max. Tube Diameter16.5 mm
Max. Tube Length121 mm
Capacity24 x 15 mL
Max. RCF3,661 x g
Max. Speed2,380 rpm
TypeFixed Angle Rotor
Unit SizeEach