1,4-Diazabicyclo[2.2.2]octane, 97%
1,4-Diazabicyclo[2.2.2]octane, 97%
1,4-Diazabicyclo[2.2.2]octane, 97%
Acros Organics

1,4-Diazabicyclo[2.2.2]octane, 97%

CAS: 280-57-9 | C6H12N2 | 112.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
112470025
also known as 11247
2.5 kgPlastic bottle
Catalog number 112470025
also known as 11247
HSN 29335990
Price (INR)
84,531.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
84,531.00
Each
Chemical Identifiers
CAS280-57-9
IUPAC Name1,4-diazabicyclo[2.2.2]octane
Molecular FormulaC6H12N2
InChI KeyIMNIMPAHZVJRPE-UHFFFAOYSA-N
SMILESC1CN2CCN1CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
GC>=96.0 %
Water=<2.0 % (K.F.)
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only