Product Image
Thermo Scientific™
Consumable Kits
Everything needed for consistent HR-ICP-MS analysis with the Thermo Scientific™ ELEMENT™ Series analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1125664Spare Parts Kit
1188700Consumable Element XR Glovebox
1188480Consumables Element XR Kit
1179480Consumables Element 2 Glovebox
1125671Consumables Element 2 Kit
Catalog number 1125664
Price (INR)
-
Type:
Spare Parts Kit
Choose these convient, Thermo Scientific Consumable Kits of bundled spares, providing everything needed for consistent, highly advanced trace elemental analysis with your Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
Select Quality Consumables for Improved Performance These useful and comprehensive kits are tailored to either the ELEMENT 2 or ELEMENT XR HR-ICP-MS analyzers to ensure you receive exactly what you need for your model of choice, from nebulizers and pump tubings, to cones, torch, and injector. even a spare Guard Electrode and load coil are included.
For the ELEMENT 2 HR-ICP-MS analyzer;
  • Consumables kit
  • Consumables kit with Glove Box
For the ELEMENT XR HR-ICP-MS analyzer;
  • Consumables kit
  • Consumables kit with Glove Box
For both the ELEMENT Series HR-ICP-MS instruments;
  • Spare parts kit, Hi-Pace Turbo-pump Version, includes replacement seals, valves, O-rings, reservoirs, oil, cones. For instruments shipped 2010 and later
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
IncludesReplacement Seals, Valves, O-Rings, Reservoirs, Oil, Cones. For Instruments Shipped 2010 and Later
TypeSpare Parts Kit
Unit SizeEach