Product Image
Thermo Scientific™
Torch Assembly
Easily exchange this complete click-in Thermo Scientific™ Torch Assembly, designed for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS.
Have Questions?
Catalog NumberType
1127110Torch Assembly
Catalog number 1127110
Price (INR)
-
Type:
Torch Assembly
Easily exchange this complete click-in Thermo Scientific™ Torch Assembly, designed for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS, that includes a guard electrode and an injector for convenient exchange.
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
TypeTorch Assembly
Unit SizeEach