1,6-Dibromohexane, 98%
1,6-Dibromohexane, 98%
1,6-Dibromohexane, 98%
Acros Organics

1,6-Dibromohexane, 98%

CAS: 629-03-8 | C6H12Br2 | 243.97 g/mol
Have Questions?
Catalog NumberQuantityPackaging
112810025
also known as 11281
2.5 LGlass bottle
Catalog number 112810025
also known as 11281
HSN 29033920
Price (INR)
71,369.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
71,369.00
Each
Chemical Identifiers
CAS629-03-8
IUPAC Name1,6-dibromohexane
Molecular FormulaC6H12Br2
InChI KeySGRHVVLXEBNBDV-UHFFFAOYSA-N
SMILESBrCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5056 to 1.5076 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only