Product Image
Thermo Scientific™

OLIGO™ R3 Reversed Phase Columns

Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions
1133226
also known as 1-1332-26
4.6 x 100 mm
1133426
also known as 1-1334-26
4.6 x 100 mm
Catalog number 1133226
also known as 1-1332-26
Price (INR)
94,589.00
Each
Dimensions:
4.6 x 100 mm
Price (INR)
94,589.00
Each

Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles.

Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.

Features of Oligo R3 resin and pre-packed columns include:

  • High performance chromatography resin for analytical to process scale separations
  • High resolution preparative purification at high flow rates
  • Chemical stability—allows aggressive cleaning and sanitization
  • Poly-divinylbenzene particles—yield a robust, easily packable matrix
  • Combines low back pressure, high pH stability, and high capacity
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column FormatPrepacked Column
Column TypeReversed Phase
Dimensions4.6 x 100 mm
Max. Pressure2400 psi (170 bar)
Quantity1 Column
Shipping ConditionRoom Temperature
Stationary PhasePhenyl
Volume (Metric)1.7 mL
Diameter (Metric)4.6 mm
Length (Metric)100 mm
MaterialPEEK
Particle Size30 μm
Pore Size350 Å
Product LinePOROS
TypeAnalytical Column
Unit SizeEach
Contents & Storage
Store at room temperature.