Product Image
Thermo Scientific™

OLIGO™ R3 Reversed Phase Columns

Thermo Scientific™ Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial
1133426
also known as 1-1334-26
Stainless Steel
1133226
also known as 1-1332-26
PEEK
Catalog number 1133426
also known as 1-1334-26
Price (INR)
81,429.00
Each
Material:
Stainless Steel
Price (INR)
81,429.00
Each

Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles.

Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.

Features of Oligo R3 resin and pre-packed columns include:

  • High performance chromatography resin for analytical to process scale separations
  • High resolution preparative purification at high flow rates
  • Chemical stability—allows aggressive cleaning and sanitization
  • Poly-divinylbenzene particles—yield a robust, easily packable matrix
  • Combines low back pressure, high pH stability, and high capacity
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column FormatPrepacked Column
Column TypeReversed Phase
Max. Pressure2400 psi (170 bar)
Product TypeAnalytical Column
Quantity1 Column
Shipping ConditionRoom Temperature
Stationary PhasePhenyl
Volume (Metric)1.7 mL
Diameter (Metric)4.6 mm
Length (Metric)100 mm
MaterialStainless Steel
Particle Size30 μm
PhaseReversed Phase
Pore Size350 Å
Product LinePOROS
TechniqueReversed Phase HPLC
Unit SizeEach