2,4-Dichlorophenoxyacetic acid, 97%
2,4-Dichlorophenoxyacetic acid, 97%
2,4-Dichlorophenoxyacetic acid, 97%
Acros Organics

2,4-Dichlorophenoxyacetic acid, 97%

CAS: 94-75-7 | C8H6Cl2O3 | 221.033 g/mol
Have Questions?
Catalog NumberQuantityPackaging
113635000
also known as 11363
500 gPlastic bottle
Catalog number 113635000
also known as 11363
HSN 29183090
Price (INR)
15,079.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
15,079.00
Each
Chemical Identifiers
CAS94-75-7
IUPAC Name2-(2,4-dichlorophenoxy)acetic acid
Molecular FormulaC8H6Cl2O3
InChI KeyOVSKIKFHRZPJSS-UHFFFAOYSA-N
SMILESOC(=O)COC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with NaOH96.0 to 104.0 %
Melting point136°C to 141°C
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only