1,2-Dichloropropane, 98%, pure
1,2-Dichloropropane, 98%, pure
1,2-Dichloropropane, 98%, pure
Acros Organics

1,2-Dichloropropane, 98%, pure

CAS: 78-87-5 | C3H6Cl2 | 112.98 g/mol
Have Questions?
Catalog NumberQuantityPackaging
113670500
also known as 11367
50 mLGlass bottle
Catalog number 113670500
also known as 11367
HSN 29031990
Price (INR)
4,267.00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,267.00
Each
Chemical Identifiers
CAS78-87-5
IUPAC Name1,2-dichloropropane
Molecular FormulaC3H6Cl2
InChI KeyKNKRKFALVUDBJE-UHFFFAOYNA-N
SMILESCC(Cl)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only