Diethyldithiocarbamic acid, silver salt, 99%, for analysis
Diethyldithiocarbamic acid, silver salt, 99%, for analysis
Diethyldithiocarbamic acid, silver salt, 99%, for analysis
Acros Organics

Diethyldithiocarbamic acid, silver salt, 99%, for analysis

CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
11429025025 gGlass bottle
Catalog number 114290250
HSN 28432900
Price (INR)
46,846.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
46,846.00
Each
Chemical Identifiers
CAS1470-61-7
IUPAC Namesilver(1+) (diethylcarbamothioyl)sulfanide
Molecular FormulaC5H10AgNS2
InChI KeyNSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES[Ag+].CCN(CC)C([S-])=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to khaki
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NH4SCN98.5 to 101.5 %
Solubility(0.5% in pyridine) clear
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only