4,8-Dihydroxyquinoline-2-carboxylic acid, 96%
4,8-Dihydroxyquinoline-2-carboxylic acid, 96%
4,8-Dihydroxyquinoline-2-carboxylic acid, 96%
Acros Organics

4,8-Dihydroxyquinoline-2-carboxylic acid, 96%

CAS: 59-00-7 | C10H7NO4 | 205.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
115090010
also known as 11509
1 gGlass bottle
Catalog number 115090010
also known as 11509
HSN 29334900
Price (INR)
8,427.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,427.00
Each
Chemical Identifiers
CAS59-00-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Ochre to dark green
Appearance (Form)Powder
Infrared spectrumConforms
Titration excess NaOH95 to 105 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only