2,5-Dimethoxybenzaldehyde, 97%
2,5-Dimethoxybenzaldehyde, 97%
2,5-Dimethoxybenzaldehyde, 97%
Acros Organics

2,5-Dimethoxybenzaldehyde, 97%

CAS: 93-02-7 | C9H10O3 | 166.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
115391000100 gGlass bottle
Catalog number 115391000
HSN 29124999
Price (INR)
12,143.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,143.00
Each
Chemical Identifiers
CAS93-02-7
IUPAC Name2,5-dimethoxybenzaldehyde
Molecular FormulaC9H10O3
InChI KeyAFUKNJHPZAVHGQ-UHFFFAOYSA-N
SMILESCOC1=CC=C(OC)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point46°C to 51°C
GC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Yellow to beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only