3,4-Dimethoxybenzoic acid, 99+%
3,4-Dimethoxybenzoic acid, 99+%
3,4-Dimethoxybenzoic acid, 99+%
Acros Organics

3,4-Dimethoxybenzoic acid, 99+%

CAS: 93-07-2 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 115455000
also known as 11545
HSN 29183090
Price (INR)
36,839.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
36,839.00
Each
Chemical Identifiers
CAS93-07-2
IUPAC Name3,4-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyDAUAQNGYDSHRET-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1OC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to beige
Infrared spectrumConforms
Melting point180°C to 184°C
HPLC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only