Dimethoxymethane, 99.5+%
Dimethoxymethane, 99.5+%
Dimethoxymethane, 99.5+%
Acros Organics

Dimethoxymethane, 99.5+%

CAS: 109-87-5 | C3H8O2 | 76.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
115560250
also known as 11556
25 LMetal drum
Catalog number 115560250
also known as 11556
HSN 29110010
Price (INR)
79,039.00
Each
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (INR)
79,039.00
Each
Chemical Identifiers
CAS109-87-5
IUPAC Namedimethoxymethane
Molecular FormulaC3H8O2
InChI KeyNKDDWNXOKDWJAK-UHFFFAOYSA-N
SMILESCOCOC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.50 %
Infrared spectrumConforms
Water=<0.03 %
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only