2,2-Dimethoxypropane, 98+%
2,2-Dimethoxypropane, 98+%
2,2-Dimethoxypropane, 98+%
Acros Organics

2,2-Dimethoxypropane, 98+%

CAS: 77-76-9 | C5H12O2 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 115630025
also known as 11563
HSN 29101000
Price (INR)
23,222.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
23,222.00
Each
Chemical Identifiers
CAS77-76-9
IUPAC Name2,2-dimethoxypropane
Molecular FormulaC5H12O2
InChI KeyHEWZVZIVELJPQZ-UHFFFAOYSA-N
SMILESCOC(C)(C)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3770 to 1.3790 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only