Spray Chamber Accessory
Actual product may vary
Thermo Scientific™
Spray Chamber Accessory
Improve analysis by matching the spray chamber to your analytes when using the advanced Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
1157350PFA Spray Chamber
1120900Quartz Spray Chamber
1127121HPM PFA Spray Chamber
1161860Borosil Cyclonic Spray Chamber
Catalog number 1157350
Price (INR)
-
Includes:
PFA Spray Chamber
Improve your analysis by matching the Thermo Scientific™ Spray Chamber to your analysis requirements when using the advanced Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
pHR-ICP-MS Spray Chambers are available in quartz or PFA, double-pass or cyclonic or even dedicated solely to the high precision measurement of isotope rations so you can tailor your spray chamber accessory to your chosen analyte precisely.
Enhance Analysis with Tailored Accessories
Select the right Spray Chamber for your analysis:
  • Spray chamber for high precision measurements (isotope ratios); recommended for use with PFA nebulizers of 100μl/min uptake rate
  • Quartz double pass spray chamber for Peltier Device; a multipurpose spray chamber for high-flow nebulizers
  • Quartz spray chamber; a multipurpose spray chamber for high-flow nebulizers
  • Borosilicate Cyclonic spray chamber (Twinnabar); a multipurpose spray chamber for nebulizer flow rates up to 400μl/min, with excellent washout characteristics and matrix tolerance. Recommended for use with the MicroMist Nebulizer 200μl/min
  • PFA spray chamber (HF resistant); for use with the Peltier Cooler and with PFA nebulizers. Fits to the 1.8mm I.D. HF resistant torch injectors in sapphire and platinum
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS system
IncludesPFA Spray Chamber
TypeSpray Chamber
Unit SizeEach