Product Image
Thermo Scientific™
HPLC Connection Kit
Configure an HPLC system to the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS instrument with the parts included in this handy Thermo Scientific™ HPLC Connection Kit.
Have Questions?
Catalog NumberTypeFor Use With (Equipment)
1159340HPLC Connection KitELEMENT Series HR-ICP-MS System
Catalog number 1159340
Price (INR)
-
Type:
HPLC Connection Kit
For Use With (Equipment):
ELEMENT Series HR-ICP-MS System
Enable the quick and easy coupling of a High Performance Liquid Chromatography (HPLC) system to the advanced Thermo Scientific™ ELEMENT™ Series HR-ICP-MS analyzer for elemental speciation analysis with the Thermo Scientific™ HPLC Connection Kit.
Everything required to configure an HPLC system to the ELEMENT Series HR-ICP-MS instrument is included in this useful HPLC Connection Kit, including a nebulizer for standard and low flow rate.
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
TypeHPLC Connection Kit
Unit SizeEach