Sample Handling Kits for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Actual product may vary
Thermo Scientific™

Sample Handling Kits for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems

Get accurate ICP-MS results using dedicated sample handling kits for the Thermo Scientific™ Element™ Series HR-ICP-MS and Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
1159440Matrix KitElement Series HR-ICP-MS
1067531Organic Matrix AccessoryElement Series HR-ICP-MS
1162170Acid-Resistant Sample IntroElement Series HR-ICP-MS
1324711Organic Matrix KitiCAP MX and iCAP Q/Qnova
1324720Acid Resistant Kit Q/QnovaiCAP Q/Qnova ICP-MS
BRE0064559iCAP MX Acid Resistance KitiCAP MX Series ICP-MS
Catalog number 1159440
Price (INR)
-
Type:
Matrix Kit
For Use With (Equipment):
Element Series HR-ICP-MS

Enhance your analysis with dedicated Thermo Scientific™ Sample Handling Kits for the Element Series HR-ICP-MS, the iCAP MX and iCAP Q/Qnova Series ICP-MS instruments.

Enable the routine analysis of organic solvents, aggressively acidic or highly complex sample matrices with these convenient, complete sample handling accessory kits.

Enhance System Performance with Tailored Sampling Kits

Organic Matrix Accessory Kit for Element Series HR-ICP-MS

Organic solvents are easily handled with this kit comprising:

  • Mass flow controller for use with oxygen
  • Dual spray chamber
  • Skimmer and sample cones (platinum tipped) not included
  • Nebulizer and injector
  • Tubing and connector

Acid-Resistant Sample Introduction System

For the routine analysis of aggressive sample matrices, this kit includes:

  • Mass flow controller for use with argon gas
  • Special pumping system with synthetic oil
  • Sampler and skimmer cones (platinum tipped) included

Complex Matrix Sample Handling Kit for Element Series HR-ICP-MS

Enable the routine analysis of highly complex matrices with this product bundle containing:

  • MicroMist nebulizer
  • Twinnabar cyclonic spray chamber
  • Injectors 2.2 mm
  • Tubing and connectors
Specifications
For Use With (Equipment)Element Series HR-ICP-MS
TypeMatrix Kit
Unit SizeEach