N,N'-Dimethylpiperazine, 98.5%
N,N'-Dimethylpiperazine, 98.5%
N,N'-Dimethylpiperazine, 98.5%
Acros Organics

N,N'-Dimethylpiperazine, 98.5%

CAS: 106-58-1 | C6H14N2 | 114.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
116650050
also known as 11665
5 mLGlass bottle
Catalog number 116650050
also known as 11665
HSN 29335990
Price (INR)
3,172.00
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,172.00
Each
Chemical Identifiers
CAS106-58-1
IUPAC Name1,4-dimethylpiperazine
Molecular FormulaC6H14N2
InChI KeyRXYPXQSKLGGKOL-UHFFFAOYSA-N
SMILESCN1CCN(C)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4453 to 1.4473 (20°C, 589 nm)
Water=<0.3 %
Infrared spectrumConforms
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only