9,10-Diphenylanthracene, 98%
9,10-Diphenylanthracene, 98%
9,10-Diphenylanthracene, 98%
Acros Organics

9,10-Diphenylanthracene, 98%

CAS: 1499-10-1 | C26H18 | 330.43 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1172000505 gGlass bottle
Catalog number 117200050
HSN 29029090
Price (INR)
22,133.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
22,133.00
Each
Chemical Identifiers
CAS1499-10-1
IUPAC Name9,10-diphenylanthracene
Molecular FormulaC26H18
InChI KeyFCNCGHJSNVOIKE-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Melting point248°C to 252°C
Appearance (Color)Light yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only