trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
trans,trans-1,4-Diphenyl-1,3-butadiene, 99%
Acros Organics

trans,trans-1,4-Diphenyl-1,3-butadiene, 99%

CAS: 538-81-8 | C16H14 | 206.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 117221000
also known as 11722
HSN 29029090
Price (INR)
58,208.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
58,208.00
Each
Chemical Identifiers
CAS538-81-8
IUPAC Name[(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
Molecular FormulaC16H14
InChI KeyJFLKFZNIIQFQBS-FNCQTZNRSA-N
SMILESC(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point151°C to 154°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only