3,3-Diphenylpropionic acid, 97%
3,3-Diphenylpropionic acid, 97%
3,3-Diphenylpropionic acid, 97%
Acros Organics

3,3-Diphenylpropionic acid, 97%

CAS: 606-83-7 | C15H14O2 | 226.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
117420250
also known as 11742
25 gGlass bottle
Catalog number 117420250
also known as 11742
HSN 29163990
Price (INR)
5,468.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,468.00
Each
Chemical Identifiers
CAS606-83-7
IUPAC Name3,3-diphenylpropanoic acid
Molecular FormulaC15H14O2
InChI KeyBZQGAPWJKAYCHR-UHFFFAOYSA-N
SMILESOC(=O)CC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point150°C to 155°C
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only