Ellagic acid, 97%
Ellagic acid, 97%
Ellagic acid, 97%
Acros Organics

Ellagic acid, 97%

CAS: 476-66-4 | C14H6O8 | 302.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
50 g
Packaging:
Glass bottle
Catalog number 117740100
HSN 29322090
Price (INR)
20,381.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
20,381.00
Each
Chemical Identifiers
CAS476-66-4
IUPAC Name6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
Molecular FormulaC14H6O8
InChI KeyAFSDNFLWKVMVRB-UHFFFAOYSA-N
SMILESOC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Beige to green
Infrared spectrumConforms
Titration with TBAH96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only