Product Image
Invitrogen™
Gateway™ BP Clonase™ Enzyme Mix
Gateway BP Clonase® enzyme contains both Int (Integrase) and IHF (Integration Host Factor) proteins that catalyze the in vitro recombinationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1178901320 Reactions
11789021100 Reactions
Catalog number 11789013
Price (INR)
51,939.00
線上優惠
Ends: 31-Mar-2026
86,565.00
Save 34,626.00 (40%)
Each
Quantity:
20 Reactions
Price (INR)
51,939.00
線上優惠
Ends: 31-Mar-2026
86,565.00
Save 34,626.00 (40%)
Each
Gateway BP Clonase® enzyme contains both Int (Integrase) and IHF (Integration Host Factor) proteins that catalyze the in vitro recombination of PCR products or DNA segments from clones (containing attB sites) and a Donor vector (containing attP sites) to generate Entry clones. BP Clonase® II enzyme format is a single mix of both enzyme and buffer to ensure enzyme stability and enable convenient 10μl reaction set up with fewer pipetting steps, which both maximize recombination efficiency and minimize pippetting. The original format of BP Clonase enzyme is the stand alone enzyme blend and reaction buffer provided in separate tubes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferReaction Buffer
Product TypeBP Clonase Enzyme Mix
Quantity20 Reactions
Shipping ConditionDry Ice
EnzymeBP Clonase
Product LineClonase, Gateway
Unit SizeEach
Contents & Storage
All Gateway™ BP Clonase™ enzyme kits include proteinase K solution (2 μg/μl), 30% PEG 8000/30 mM MgCl2 Solution and a positive control vector. Store BP Clonase™ enzyme at -80°C; BP Clonase™ II Enzyme mixtures at -20°C. Guaranteed stable for 6 months when properly stored.
Frequently asked questions (FAQs)
I performed a BP reaction and got high background after transformation. Can you please offer some troubleshooting tips?
– Check whether the reaction was transformed into an E.coli strain containing the F' episome and the ccdA gene – use an E.coli strain that does not contain the F' episome, e.g. DH10B, TOP10.
– Deletion (full or partial) of the ccdB gene – propagate in media with 50-100 mg/mL ampicillin and 15-30 µg/mL chloramphenicol.
– Contamination from another resistant strain.
– Check whether proper amount of DNA was used in the reaction.
I performed a BP reaction and got two distinct types of colonies (large and small) after transformation. How should I proceed?
Typically when both large and small colonies are produced following BP recombination, we recommend screening several of both of the small and large colony types by analytical restriction digest, PCR with gene specific primers, or by sequencing to determine what you have and decide on the best course forward. If one of the colonies you analyze contains your desired entry clone, then you may proceed with this entry clone to an LR reaction to produce your desired expression clone.
There are several possible causes of this issue:
– Plasmid was lost during culture due to large size or toxicity – To improve results next time you perform a BP reaction with a tricky insert, you may try incubating your transformation plate at 30 degrees C and/or use Stbl2 E.coli to stabilize the plasmid. We also recommend performing the BP reaction positive control pEXP7-Tet alongside your BP cloning reaction of interest; the results of that control reaction will let you know whether the large and small colony phenotype is specific to your insert of interest.
– Deletions (full or partial) or point mutations in the ccdB gene – A negative BP reaction control (with no BP clonase added) should not produce any colonies – If a no BP clonase negative control produces colonies, then the ccdB/chlorophenicol cassette is compromised and we recommend to obtain a new pDONR vector.br/>
– Background on antibiotic selection plate due to contamination or expired antibiotic – If you run a no plasmid added transformation negative control plate, then this plate should not produce any colonies. Colonies on transformation negative control plate suggests contamination or need to make fresh plates.
I performed a BP reaction and got no colonies after transformation, and the recombination positive control was not successful. Can you please offer some suggestions?
– Check the competent cells with pUC19 transformation.
– Increase the amount plated.
I performed a BP reaction and got few or no colonies after transformation, whereas the transformation control gave colonies. Can you please offer some suggestions?
– Increase the incubation time up to 18 hours.
– Make sure to treat reactions with proteinase K before transformation.
– Check whether the correct antibiotic was used for selection.
– Check whether the att site sequences are correct.
– Check whether the correct Clonase enzyme was used and whether it was functional.
– Check whether the recommended amount of DNA was used in the reaction.
– Check primer design and try gel/PEG purifying the attB-PCR product.
– If the attB-PCR product or linear attB Expression clone is too long (>5 kb), incubate the BP reaction overnight.
What is the difference between BP Clonase and BP Clonase II and between LR Clonase and LR Clonase II?
BP Clonase and LR Clonase have the 5X BP reaction buffer or the 5X LR reaction buffer, respectively, in separate vials and require storage at -80 degrees C. On the other hand, BP Clonase II and LR Clonase II offer the convenience of pre-mixed BP Clonase /LR Clonase and 5X BP/LR reaction buffer, respectively. These pre-mixed formulations are more stable at -20 degrees C.
Citations & References (13)
Citations & References
Abstract
A suite of Gateway cloning vectors for high-throughput genetic analysis in Saccharomyces cerevisiae.
Authors:Alberti S, Gitler AD, Lindquist S,
Journal:Yeast
PubMed ID:17583893
'In the post-genomic era, academic and biotechnological research is increasingly shifting its attention from single proteins to the analysis of complex protein networks. This change in experimental design requires the use of simple and experimentally tractable organisms, such as the unicellular eukaryote Saccharomyces cerevisiae, and a range of new high-throughput ... More
The alliance for cellular signaling plasmid collection: a flexible resource for protein localization studies and signaling pathway analysis.
Authors:Zavzavadjian JR, Couture S, Park WS, Whalen J, Lyon S, Lee G, Fung E, Mi Q, Liu J, Wall E, Santat L, Dhandapani K, Kivork C, Driver A, Zhu X, Chang MS, Randhawa B, Gehrig E, Bryan H, Verghese M, Maer A, Saunders B, Ning Y, Subramaniam S, Meyer T, Simon MI, O'Rourke N, Chandy G, Fraser ID,
Journal:Mol Cell Proteomics
PubMed ID:17192258
'Cellular responses to inputs that vary both temporally and spatially are determined by complex relationships between the components of cell signaling networks. Analysis of these relationships requires access to a wide range of experimental reagents and techniques, including the ability to express the protein components of the model cells in ... More
Gateway RFP-fusion vectors for high throughput functional analysis of genes.
Authors:Park JY, Hwang EM, Park N, Kim E, Kim DG, Kang D, Han J, Choi WS, Ryu PD, Hong SG,
Journal:Mol Cells
PubMed ID:17646710
'There is an increasing demand for high throughput (HTP) methods for gene analysis on a genome-wide scale. However, the current repertoire of HTP detection methodologies allows only a limited range of cellular phenotypes to be studied. We have constructed two HTP-optimized expression vectors generated from the red fluorescent reporter protein ... More
High-throughput yeast two-hybrid assays for large-scale protein interaction mapping.
Authors:Walhout AJ, Vidal M,
Journal:Methods
PubMed ID:11403578
'Protein-protein interactions play fundamental roles in many biological processes. Hence, protein interaction mapping is becoming a well-established functional genomics approach to generate functional annotations for predicted proteins that so far have remained uncharacterized. The yeast two-hybrid system is currently one of the most standardized protein interaction mapping techniques. Here, we ... More
Towards a proteome-scale map of the human protein-protein interaction network.
Authors:Rual JF, Venkatesan K, Hao T, Hirozane-Kishikawa T, Dricot A, Li N, Berriz GF, Gibbons FD, Dreze M, Ayivi-Guedehoussou N, Klitgord N, Simon C, Boxem M, Milstein S, Rosenberg J, Goldberg DS, Zhang LV, Wong SL, Franklin G, Li S, Albala JS, Lim J, Fraughton C, Llamosas E, Cevik S, Bex C, Lamesch P, Sikorski RS, Vandenhaute J, Zoghbi HY, Smolyar A, Bosak S, Sequerra R, Doucette-Stamm L, Cusick ME, Hill DE, Roth FP, Vidal M,
Journal:Nature
PubMed ID:16189514
'Systematic mapping of protein-protein interactions, or ''interactome'' mapping, was initiated in model organisms, starting with defined biological processes and then expanding to the scale of the proteome. Although far from complete, such maps have revealed global topological and dynamic features of interactome networks that relate to known biological properties, suggesting ... More