Everything needed for consistent HR-ICP-MS analysis with the Thermo Scientific™ ELEMENT™ Series analyzer.
Have Questions?
Change view
Catalog Number
Type
1179480
Consumables Element 2 Glovebox
1188700
Consumable Element XR Glovebox
1188480
Consumables Element XR Kit
1125671
Consumables Element 2 Kit
1125664
Spare Parts Kit
5 Options
Catalog number 1179480
Price (INR)
-
Type:
Consumables Element 2 Glovebox
Choose these convient, Thermo Scientific Consumable Kits of bundled spares, providing everything needed for consistent, highly advanced trace elemental analysis with your Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
Select Quality Consumables for Improved Performance These useful and comprehensive kits are tailored to either the ELEMENT 2 or ELEMENT XR HR-ICP-MS analyzers to ensure you receive exactly what you need for your model of choice, from nebulizers and pump tubings, to cones, torch, and injector. even a spare Guard Electrode and load coil are included.
For the ELEMENT 2 HR-ICP-MS analyzer;
Consumables kit
Consumables kit with Glove Box
For the ELEMENT XR HR-ICP-MS analyzer;
Consumables kit
Consumables kit with Glove Box
For both the ELEMENT Series HR-ICP-MS instruments;
Spare parts kit, Hi-Pace Turbo-pump Version, includes replacement seals, valves, O-rings, reservoirs, oil, cones. For instruments shipped 2010 and later
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS system
IncludesNebulizers and Pump Tubings, to Cones, Torch, Injector, Spare Guard Electrode, and Load Coil