Lactalbumin Hydrolysate, powder (extra soluble)
Product Image
Gibco™

Lactalbumin Hydrolysate, powder (extra soluble)

Tissue culture grade.Read more
Have Questions?
Catalog NumberQuantity
118000425 kg
Catalog number 11800042
Price (INR)
81,828.00
Each
Add to cart
Quantity:
5 kg
Price (INR)
81,828.00
Each
Add to cart
Tissue culture grade.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Quantity5 kg
Shipping ConditionRoom Temperature
FormPowder
Product TypeLactalbumin Hydrolysate
Unit SizeEach
Contents & Storage
Store at room temperature.