Take advantage of technology improvements with these in-field Upgrade Kits for the Thermo Scientific™ ELEMENT™ HR-ICP-MS analyzer.
Have Questions?
Change view
Catalog Number
Includes
1180080
SEM ICP2-V2 Upgrade Kit
1156420
Peltier Cooling ICP
1179730
SEV ICP2-V2 PLUG IN Upgrade
1181030
Retrofit-Kit for Plug-in Slit
1289760
Pump for Jet Interface
5 Options
Catalog number 1180080
Price (INR)
-
Includes:
SEM ICP2-V2 Upgrade Kit
Enhance your Thermo Scientific™ ELEMENT™ HR-ICP-MS analyzer with these in-field retrofit Thermo Scientific™ Upgrade Kits to enable your HR-ICP-MS analysis capabilities to improve and progress in line with advances in technology.
Upgrade Kits for Improved Performance
Multiplier SEM Type II Detector; two specific retrofit kit versions, for either pre- or post-February 2003 instrument shipments
Peltier cooling for spray chamber; in-field retrofit kit
Plug-in slit kit; retrofit kit for ELEMENT 2 HR-ICP-MS model for the user-replaceable slit tongues
High capacity Interface Pump for Jet Interface; in-field upgrade
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS system