N-Ethylaniline, 98%
N-Ethylaniline, 98%
N-Ethylaniline, 98%
Acros Organics

N-Ethylaniline, 98%

CAS: 103-69-5 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number 118045000
HSN 29214290
Price (INR)
12,096.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,096.00
Each
Chemical Identifiers
CAS103-69-5
IUPAC NameN-ethylaniline
Molecular FormulaC8H11N
InChI KeyOJGMBLNIHDZDGS-UHFFFAOYSA-N
SMILESCCNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow to light brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5530 to 1.5560 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only