Flat Stainless Steel Bath Covers for Lindberg/Blue M™ Baths
Thermo Scientific™
Flat Stainless Steel Bath Covers for Lindberg/Blue M™ Baths
Minimize heat loss and maintain bath temperature until ready for next application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
118081Water Bath Models WB1110
118082Water Bath Models WB1120
118083Water Bath Models WB1130
118084WB1140
118085SWB1122, RSWB3222
Catalog number 118081
Price (INR)
-
For Use With (Equipment):
Water Bath Models WB1110
Minimize heat loss and maintain bath temperature until ready for next application with Thermo Scientific™ Flat Stainless-Steel Covers.
  • Protect the medium when bath is not in use
  • Covers include handles
  • Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)Water Bath Models WB1110
TypeCover
Unit SizeEach