Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Acros Organics

Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 118450010
HSN 29094400
Price (INR)
22,519.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
22,519.00
Each
Chemical Identifiers
CAS110-71-4
IUPAC Name1,2-dimethoxyethane
Molecular FormulaC4H10O2
InChI KeyXTHFKEDIFFGKHM-UHFFFAOYSA-N
SMILESCOCCOC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3785 to 1.3815 (20°C, 589 nm)
Water=<0.1 % (K.F.)
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only