Product Image
Thermo Scientific™
Sample Racks for Lindberg/Blue M™ Shaking Circulating Water Baths
Use with SWB1122 and RSWB3222 Lindberg/Blue M Circulating Water Baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
11846625 mm Tubes
11807725 x 25 mL or 23 x 50 mL Flasks
118078125 mL, 250 mL, 500 mL, 1L Flasks
11846513 mm Tubes
Catalog number 118466
Price (INR)
-
Holds:
25 mm Tubes
The Thermo Scientific™ Sample Racks are for use with Thermo Scientific™ SWB1122 and RSWB3222 Lindberg/Blue M Circulating Water Baths.
Specifications
DescriptionTube Rack
For Use With (Equipment)Water Bath Models SWB1122, RSWB3222
Holds25 mm Tubes
TypeTube Rack
Unit SizeEach