4-Ethylphenol, 97%
4-Ethylphenol, 97%
4-Ethylphenol, 97%
Acros Organics

4-Ethylphenol, 97%

CAS: 123-07-9 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 118820050
also known as 11882
HSN 29071990
Price (INR)
3,503.00
Each
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
3,503.00
Each
Chemical Identifiers
CAS123-07-9
IUPAC Name4-ethylphenol
Molecular FormulaC8H10O
InChI KeyHXDOZKJGKXYMEW-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Infrared spectrumConforms
Appearance (Form)Crystals or crystalline solid or fused mass
GC>=96.0 %
Melting point38°C to 45°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only