Product Image
Thermo Scientific™
Additional Mass Flow Controllers
Gain increased flexibility from your Thermo Scientific™ ELEMENT™ Series HR-ICP-MS instrument with these Thermo Scientific™ Additional Mass Flow Controllers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1189230Mass Flow Controller (Helium)
1123900Mass Flow Controller (Argon)
1123910Mass Flow Controller (Oxygen)
1271870Gas MFC Kit (1000mL/min)
1322530Gas MFC Kit (250mL/min)
Catalog number 1189230
Price (INR)
-
Type:
Mass Flow Controller (Helium)
Gain increased flexibility from your Thermo Scientific™ ELEMENT™ Series HR-ICP-MS instrument with these ThermoScientific™ Additional Mass Flow Controllers (MFCs), to enable the introduction of make-up-gases, argon, oxygen or helium, into the spray chamber or to supply these gases for advanced hyphenated analysis techniques. Two additional MFCs can be installed simultaneously.
Increase Flexibility with Customized Accessories
  • Mass flow controller for addition of make-up-gas (argon), 1NL/min; for spray-chambers with additional gas port
  • Mass flow controller for addition of make- up-gas (oxygen), 1NL/min; for organic matrix analysis
  • Mass flow controller for addition of make-up-gas (helium), 2NL/min; for Laser Ablation analysis
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
TypeMass Flow Controller (Helium)
Unit SizeEach