Product Image
Thermo Scientific™
Lindberg/Blue M™ Vacuum Oven Accessories
Add Thermo Scientific Lindberg Blue M Vacuum Ovens Accessories to your vacuum oven for a more effective lab tool.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
118961Connection KitLindberg/Blue M Vacuum Ovens
118974Buna-N Door gasketVO914 Series
118977Buna-N Door gasketVO1218 Series
305992H01Silicone (red) Door gasketVO1218 Series
305993H01Silicone (red) Door gasketVO1824 Series
305994H01Silicone (red) Door gasketVO914 Series
34637H01Buna-N Door gasketVO1824 Series
3497TS
also known as 3497
HEPA FilterLindberg/Blue M Vacuum Ovens
6718White Floor StandLindberg/Blue M Vacuum Ovens
6723Stainless-steel Floor StandLindberg/Blue M Vacuum Ovens
6831Vacuum Pump OilVacuum Pumps
6832Mist Filter ElementVacuum Pumps
6834Activated AluminaLindberg/Blue M Vacuum Ovens
Catalog number 118961
Price (INR)
-
Type:
Connection Kit
For Use With (Equipment):
Lindberg/Blue M Vacuum Ovens
Configure Thermo Scientific™ Lindberg/Blue M™ Vacuum Oven Accessories with your vacuum oven for a more effective lab tool.
Specifications
IncludesCentering Rings, Connectors, Hose to Connect Vacuum Pump, NW Fittings, Vacuum Clamps
For Use With (Equipment)Lindberg/Blue M Vacuum Ovens
TypeConnection Kit
Unit SizeEach