Sample Holders
Actual product may vary
Thermo Scientific™
Sample Holders
Get a more convenient, straightforward process with these Thermo Scientific™ Sample Holders, designed specifically for the Thermo Scientific™ ELEMENT GD™ GD-MS instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1190550Sample Holder, Standard
1178090Sample Holder,Thin Sample
1184170Rear Side Cooling, Thin Sample
Catalog number 1190550
Price (INR)
-
Type:
Sample Holder, Standard
Get a more convenient, straightforward process with these Thermo Scientific™ Sample Holders, designed specifically for the Thermo Scientific™ ELEMENT GD™ GD-MS instrument. Even the most sophisticated elemental analysis, such as Glow Discharge Mass Spectrometry (GD-MS) is made more straightforward with the right accessories. These convenient Sample Holders are designed specifically for the ELEMENT GD GD-MS instrument, with options for standard or thin samples.
Gain Simplicity from Convenient Accessories
  • Sample holder featuring an ingenious clamping mechanism. For sample diameters up to 70mm, and maximum thickness of 40mm
  • Sample holder for thin samples, for use with rear side cooling; improves temperature stability of tiny samples. Maximum sample dimensions of 45 x 45mm and minimal thickness ∼0.1mm, depending on sample material
  • Rear-side cooling, for use with the sample holder for thin samples
Specifications
For Use With (Equipment)ELEMENT GD GD-MS Instrument
TypeSample Holder, Standard
Unit SizeEach