9-Fluorenone, 99+%
9-Fluorenone, 99+%
9-Fluorenone, 99+%
Acros Organics

9-Fluorenone, 99+%

CAS: 486-25-9 | C13H8O | 180.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 119201000
also known as 11920
HSN 29143990
Price (INR)
5,539.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
5,539.00
Each
Chemical Identifiers
CAS486-25-9
IUPAC Name9H-fluoren-9-one
Molecular FormulaC13H8O
InChI KeyYLQWCDOCJODRMT-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Melting point81°C to 85°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only