Fluorescein, pure
Fluorescein, pure
Fluorescein, pure
Acros Organics

Fluorescein, pure

CAS: 2321-07-5 | C20H12O5 | 332.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number 119240025
also known as 11924
HSN 32042090
Price (INR)
53,024.00
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (INR)
53,024.00
Each
Chemical Identifiers
CAS2321-07-5
IUPAC Name3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC20H12O5
InChI KeyGNBHRKFJIUUOQI-UHFFFAOYSA-N
SMILESOC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-brown to rust brown to brown
Infrared spectrumConforms
Specific absorbance A (1%/1cm)>=1800 (490 +/- 5 nm , 0.5% aq. soln. of Na2CO3)
Appearance (Form)Powder
Loss on drying=<3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only