Pin Sample Kit
Thermo Scientific™
Pin Sample Kit
Enable the trace metal analysis of ultra-thin rods with millimeter diameters with this Thermo Scientific™ Pin Sample Kit for the Thermo Scientific™ ELEMENT™ GD GD-MS analyzer.
Have Questions?
Catalog NumberType
1195130Pin Sample Kit
Catalog number 1195130
Price (INR)
-
Type:
Pin Sample Kit
Enable the trace metal analysis of thin rods with diameters ranging from 1mm to 3mm with this Thermo Scientific™ Pin Sample Kit for the Thermo Scientific™ ELEMENT GD™ GD-MS analyzer.
Increase Analysis Capabilities with Advanced Accessories
  • Pin Sample Kit for round pins of 1-3mm diameter, 10-25mm length. Includes a dedicated sampler holder and stainless steel consumables
  • Smaller diameter Pin Sample Kits are available upon request
Specifications
For Use With (Equipment)ELEMENT GD GD-MS Instrument
TypePin Sample Kit
Unit SizeEach