Anode Tube
Thermo Scientific™
Anode Tube
Use only this superior quality replacement Anode Tube to ensure consistently high performance from your Thermo Scientific™ ELEMENT™ GD GD-MS spectrometer.
Have Questions?
Catalog NumberType
1195180Anode Tube
Catalog number 1195180
Price (INR)
-
Type:
Anode Tube
Use only this superior quality replacement Thermo Scientific™ Anode Tube to ensure consistently high performance from your Thermo Scientific™ ELEMENT GD™ GD-MS spectrometer. The durable, stainless steel anode tube comes complete with three O-rings for easy installation. All types of anode caps and flow tubes can be used with this anode tube.
Specifications
For Use With (Equipment)ELEMENT GD GD-MS Instrument
TypeAnode Tube
Unit SizeEach