4-Fluorophenol, 99%
4-Fluorophenol, 99%
4-Fluorophenol, 99%
Acros Organics

4-Fluorophenol, 99%

CAS: 371-41-5 | C6H5FO | 112.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
119560250
also known as 11956
25 gGlass bottle
Catalog number 119560250
also known as 11956
HSN 29081900
Price (INR)
5,894.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,894.00
Each
Chemical Identifiers
CAS371-41-5
IUPAC Name4-fluorophenol
Molecular FormulaC6H5FO
InChI KeyRHMPLDJJXGPMEX-UHFFFAOYSA-N
SMILESOC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline solid
Appearance (Color)Yellow to beige to pink
Melting point44°C to 49°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only