Gluconic acid, 50 wt% solution in water
Gluconic acid, 50 wt% solution in water
Gluconic acid, 50 wt% solution in water
Acros Organics

Gluconic acid, 50 wt% solution in water

CAS: 526-95-4 | C6H12O7 | 196.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 119890025
also known as 11989
HSN 29181690
Price (INR)
8,309.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,309.00
Each
Chemical Identifiers
CAS526-95-4
IUPAC Namepotassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular FormulaC6H11KO7
InChI KeyHLCFGWHYROZGBI-JJKGCWMISA-M
SMILES[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown-yellow
Titration excess NaOH49 to 55 %
Appearance (Form)Solution
Reducing sugars=<3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only