Glutathione, 98%, for analysis, reduced
Glutathione, 98%, for analysis, reduced
Glutathione, 98%, for analysis, reduced
Acros Organics

Glutathione, 98%, for analysis, reduced

CAS: 70-18-8 | C10H17N3O6S | 307.32 g/mol
Have Questions?
Catalog NumberQuantity
120000250
also known as 12000
25 g
Catalog number 120000250
also known as 12000
HSN 29309099
Price (INR)
13,256.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
13,256.00
Each
Chemical Identifiers
CAS70-18-8
IUPAC Name2-amino-4-({1-[(carboxymethyl)carbamoyl]-2-sulfanylethyl}carbamoyl)butanoic acid
Molecular FormulaC10H17N3O6S
InChI KeyRWSXRVCMGQZWBV-UHFFFAOYNA-N
SMILESNC(CCC(=O)NC(CS)C(=O)NCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Iron (Fe)=<10 ppm
Appearance (Color)White to almost white
Infrared spectrumConforms
Sulfated ash=<0.1 % (600°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only