Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Actual product may vary
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers
Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers

Nalgene™ Polypropylene Griffin Low-Form Plastic Beakers

Carry out precise and efficient pouring with these low-form beakers, featuring a single-spout design and easy-to-read silk-screened graduations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
1201-40004000 mL
1201-003030 mL
1201-005050 mL
1201-0150150 mL
1201-0250250 mL
1201-0400400 mL
1201-0600600 mL
1201-10001000 mL
1201-20002000 mL
Catalog number 1201-4000
HSN 39269099
Price (INR)
19,025.00
Case
Capacity (Metric):
4000 mL
Price (INR)
19,025.00
Case

Carry out precise and efficient pouring with Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Beakers, featuring a single-spout design and easy-to-read silk-screened graduations. Rugged, reusable, construction that will last for years under typical lab conditions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Ribs for easy stacking
  • Silk-screened with graduations, size code, resin code, catalog number, max. temperature and No Flame symbol for easy identification and safe use
  • Autoclavable/Graduated
  • Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability

Ordering Information: Available in a size variety pack using Cat. No. 1201-1234

Recycling Information: Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene labware at end of product life in communities where polypropylene can be recycled.

Nalgene beakers and graduated cylinders are delivered in corrugate cartons made from 100% post-consumer recycled material. Packaging bags are made from LDPE and are readily recyclable once the label is removed. Please continue the cycle and recycle your cartons and bags.

Specifications
Capacity (English)135.25 oz.
Condenser TypeAbbe
ColorNatural translucent
DescriptionBeaker Griffin Low-Form PP 4000 mL
Eyepiece10X/18 mm
Head StyleRotating
Lighting TypeHalogen
Capacity (Metric)4000 mL
ModelMax Mono II
MaterialPPCO
Objective TypesAchromatic
Graduation Range500 to 4000 mL
Increments250 mL
Quantity1/Pk.; 4/Cs.
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceISO/DIN 7056
Scale500 mL to 4000 mL in 250 mL increments
StyleGriffin, Low Form
ACT LabelYes
Electrical Requirements220/240 V 50/60 Hz
Voltage220/240 V
Frequency50/60 Hz
Unit SizeCase

Frequently asked questions (FAQs)

Several of your labware products are available in polypropylene (PP, PPCO) or polymethylpentene (PMP). Which should I choose?

These two materials were chosen as labware raw materials for their durability, broad chemical compatibility profile, and wide range of use temperature. Polymethylpentene offers a higher level of transparency (comparable to glass) but is slightly more brittle and therefore somewhat more likely than polypropylene to crack if dropped. Chemical or environmental factors in some applications may strongly indicate the use of only one material- please refer to laboratory plastics reference materials or call Thermo Scientific Laboratory Products Essentials technical support for assistance.