Glycylglycine, 99+%
Glycylglycine, 99+%
Glycylglycine, 99+%
Acros Organics

Glycylglycine, 99+%

CAS: 556-50-3 | C4H8N2O3 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
100 g
Catalog number 120140010
also known as 12014
HSN 29241900
Price (INR)
73,760.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
73,760.00
Each
Chemical Identifiers
CAS556-50-3
IUPAC Namehydrogen 2-(2-aminoacetamido)acetic acid chloride
Molecular FormulaC4H9ClN2O3
InChI KeyYHBAZQDEMYQPJL-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Heavy metals (as Pb)=<5 ppm
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C, 4 h)
pH5.5 to 6.5 (5 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only