2-Heptanol, 99+%
2-Heptanol, 99+%
2-Heptanol, 99+%
Acros Organics

2-Heptanol, 99+%

CAS: 543-49-7 | C7H16O | 116.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 120370500
HSN 29051900
Price (INR)
3,953.00
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,953.00
Each
Chemical Identifiers
CAS543-49-7
IUPAC Nameheptan-2-ol
Molecular FormulaC7H16O
InChI KeyCETWDUZRCINIHU-UHFFFAOYNA-N
SMILESCCCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Specific gravity(20°C) 0.816 to 0.819
Appearance (Form)Liquid
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only